Learn More
Cytiva (formerly Pall Lab) Omega Membrane Discs
Omega Membrane Discs with high performance and stability for users of conventional, non-disposable stirred cells.
Brand:  Cytiva (formerly Pall Lab) OM030076
Additional Details : Weight : 0.22520kg
 View more versions of this product
Product Code. 17104821
  • £349.00 / Pack of 12
Estimated Shipment: 25-10-2024
to see stock.
Add to basket
Description
Description
  • Polyethersulfone membrane provides higher flow rates and lower protein binding than competitive regenerated cellulose
  • Lower processing times and the highest possible recoveries
  • Discs can be washed and reused
  • Replacement membranes for other stirred cell systems concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules
  • For research use only
Specifications
Specifications
PES
Non-sterile
76 mm
30 kDa
12/Pk.