Learn More
Cytiva (formerly Pall Lab) Omega Membrane Discs
Omega Membrane Discs with high performance and stability for users of conventional, non-disposable stirred cells.
£263.00 - £359.00
Product Code | Brand | MWCO | Diameter (Metric) Membrane | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Diameter (Metric) Membrane | Price | Quantity & Availability | ||||
17154811
|
Cytiva (formerly Pall Lab)
OM010043 |
10 kDa | 43 mm |
£263.00
Pack of 12 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
17184811
|
Cytiva (formerly Pall Lab)
OM030043 |
30 kDa | 43 mm |
£263.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
17174811
|
Cytiva (formerly Pall Lab)
OM010076 |
10 kDa | 76 mm |
£349.00
Pack of 12 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
17134811
|
Cytiva (formerly Pall Lab)
OM005043 |
5 kDa | 43 mm |
£295.00
Pack of 12 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
17104821
|
Cytiva (formerly Pall Lab)
OM030076 |
30 kDa | 76 mm |
£349.00
Pack of 12 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
17144811
|
Cytiva (formerly Pall Lab)
OM005076 |
5 kDa | 76 mm |
£349.00
Pack of 12 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
17194811
|
Cytiva (formerly Pall Lab)
OM030062 |
30 kDa | 62 mm |
£359.00
Pack of 12 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
17114811
|
Cytiva (formerly Pall Lab)
OM001043 |
1 kDa | 43 mm |
£307.00
Pack of 12 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
17124811
|
Cytiva (formerly Pall Lab)
OM001076 |
1 kDa | 76 mm |
£349.00
Pack of 12 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
17164811
|
Cytiva (formerly Pall Lab)
OM010062 |
10 kDa | 62 mm |
£320.00
Pack of 12 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
17114821
|
Cytiva (formerly Pall Lab)
OM100043 |
100 kDa | 43 mm |
£311.00
Pack of 12 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
Description
- Polyethersulfone membrane provides higher flow rates and lower protein binding than competitive regenerated cellulose
- Lower processing times and the highest possible recoveries
- Discs can be washed and reused
- Replacement membranes for other stirred cell systems concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules
- For research use only
Specifications
PES | |
Non-sterile | |
12/Pk. |