Learn More
Cytiva (formerly Pall Lab) Omega Membrane Discs
Omega Membrane Discs with high performance and stability for users of conventional, non-disposable stirred cells.
Brand: Cytiva (formerly Pall Lab) OM010062
Additional Details : Weight : 0.15900kg
Description
- Polyethersulfone membrane provides higher flow rates and lower protein binding than competitive regenerated cellulose
- Lower processing times and the highest possible recoveries
- Discs can be washed and reused
- Replacement membranes for other stirred cell systems concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules
- For research use only
Specifications
PES | |
Non-sterile | |
62 mm |
10 kDa | |
12/Pk. |