Learn More
Cytiva VIA Extractor™ tissue disaggregator - Omics Bundle
Semi-automated system for consistent, rapid disaggregation of solid tissue, including tumors, into viable single cells. The VIA Extractor includes tissue disaggregator, controlled-rate freezer and the Omics clamp.
Brand:  Cytiva 29517120
Product Code. 30057808
  • £38100.00 / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The VIA Extractor™ tissue disaggregator disaggregates solid tissue into viable single cells
  • For use in high-throughput omics and flow cytometry research
  • The closed system uses mild processing to optimize yield and preserve content relative to the parent sample
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tissue Disaggregator
≤460 rpm
24 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva VIA Extractor™ tissue disaggregator - Omics Bundle > Tissue Homogenizer
For research use only (RUO). Not for diagnostic use.