Learn More
Diba Omnifit™ O-Rings for Chromatography Columns
Select from FKM or FFKM (Chemraz™) Omnifit™ O-Rings for Chromatography Columns
Brand:  Diba 006OR-03-V
Encompass_Preferred
 View more versions of this product
Product Code. 15835564
  • POA / Pack of 6
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Specifications
Omnifit™ HiT Chromatography Columns
3 mm
3 mm
6 Pk.
Liquid Chromatography Column
FKM
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Diba Omnifit™ O-Rings for Chromatography Columns