Learn More
Diba Omnifit™ Replacement Endpieces for EZ Columns
Select from fixed or adjustable Omnifit™ Replacement Endpieces for EZ Columns
Brand:  Diba 006EAE-50
Encompass_Preferred
 View more versions of this product
Product Code. 15855264
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Specifications
Adjustable
50 mm
15 cm
Omnifit™ EZ Columns
Liquid Chromatography Column
PTFE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Diba Omnifit™ Replacement Endpieces for EZ Columns