Learn More
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Mix and match your Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Column lengths while using your existing endpieces for ultimate flexibility
Brand: Diba 006ERG-06-10
This item is not returnable.
View return policy
Description
Use a different length tube with your existing endpieces to meet your specific requirements.Specifications
Omnifit™ EZ and EZ SolventPlus™ Columns | |
Liquid Chromatography Column | |
Glass |
6.6 mm | |
100 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Spot an opportunity for improvement?Share a Content Correction