Learn More
Kolmi Hopen™ Op-Air™ One Type IIR Anti-fog, Splash-resistant Medical Mask
Splash-resistant medical mask with anti-fog nose piece designed specially for those with sensitive skin.
Marque: Kolmi Hopen™ M34301F-30
Informations supplémentaires : Weight : 1.52000kg
Description
- Prevents appearance of fogging on glasses
- Absorbs sweat and moisture for sensitive skin
- Very high breathability
Spécification
Honey | |
Polypropylene | |
Pleated | |
Yes | |
Yes |
Ties | |
175 x 95 mm | |
CE, (EU) 2017/745, EN 14683:2019+AC:2019 | |
Dental, Medical | |
40 Pk. |