Learn More
Kolmi Hopen™ Op-Air™ One Type IIR Splash-resistant Medical Mask
These splash-resistant medical masks are designed for those with sensitive skin, providing less irritation against the face.
Brand:  Kolmi Hopen™ M34301-30
Additional Details : Weight : 1.52000kg
 View more versions of this product
Product Code. 17710209
  • £4.85 / Pack of 40
Estimated Shipment: 13-08-2024
to see stock.
Add to basket
Description
Description
  • Protects environment from droplets from wearer
  • Provides protection against biological fluids
  • Effective against bacteria, particles, and viruses
  • Non-irritating inner layer
  • Inner layer is specially designed to reduce moisture and wick away sweat
  • Used by healthcare workers to reduce risk of infection or in the operating room
Specifications
Specifications
Honey
Polypropylene
(EU) 2017/745, EN 14683:2019+AC:2019, ISO 9001, ISO 13485
≥99%
Yes
Ties
Pleated
Yes
Medical
40 Pk.