Learn More
Kolmi Hopen™ Op-Air™ Type II Printed Medical Masks with Earloops
These medical masks come in unique designs and have a soft inner layer for those with sensitive skin.
Brand:  Kolmi Hopen™ M76100-GEO
Additional Details : Weight : 3.70500kg
 View more versions of this product
Product Code. 17770219
  • £5.95 / Pack of 60
Estimated Shipment: 13-08-2024
to see stock.
Add to basket
Description
Description
  • Protects environment from droplets emitted by wearer
  • Feature exclusive Origami folding and Smile welding for better comfort and perfect fit
  • Softex inner layer is soft and non-irritating for maximum comfort
  • Won't wear out, even during prolonged use
  • Intended for healthcare professionals
  • May also be worn by patients to reduce risk of spreading infection
Specifications
Specifications
Blue Geometric Print
175 x 88 mm
Yes
60 Pk.
Polypropylene
(EU) 2017/745, EN 14683:2019+AC:2019, ISO 9001, ISO 13485
Yes