Learn More
Medline Scientific™ Open-Rings Cover
Minimizes liquid loss due to evaporation.
Brand: Medline Scientific™ AAA45527
Code : NEW
Additional Details : Weight : 3.50000kg
Description
- Ring size adjusted for each glassware
- Allows beakers or Erlenmeyer flasks to be placed directly onto the perforated stainless steel half-shelf adjuster or water tank bottom
Specifications
Open-Rings Covers | |
BW-20H/BW-20B | |
Minimizing evaporation loss | |
6 openings of 110mm dia. |