missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ Open-Top Tri-coded Tubes
GREENER_CHOICE

Securely store and track samples in Thermo Scientific™ Matrix™ Open-Top Tri-coded tubes. Tubes feature permanent, high contrast 2D barcodes on the bottom, plus 1D linear barcodes with human readable text on the side.

£806.00 - £924.00

Specifications

Product Type Storage Tube (Open Top)
Autoclavable Not Autoclavable
Product Line Matrix™
Graduated No
View More Specs

Products 3
Product Code Brand Capacity (Metric) Sterility Packaging Price Quantity & Availability  
Product Code Brand Capacity (Metric) Sterility Packaging Price Quantity & Availability  
17278820
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3790-WP1D
1400 μL Non-sterile Bulk
£806.00
Pack of 1000
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
 
17288820
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3791-WP1D-BR
1400 μL Non-sterile 10 Latch Racks of 96 Tubes/Cs.
£924.00
Pack of 480
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
 
17298820
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3792-WP1D-BR
1400 μL Sterile 10 Latch Racks of 96 Tubes/Cs.
£918.00
Pack of 480
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
 
Description
Show More
Specifications

Specifications

Storage Tube (Open Top)
Not Autoclavable
Matrix™
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Matrix™ Open-Top Tri-coded Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.