missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Opti-MEM™ I Reduced Serum Medium
GREENER_CHOICE

Opti-MEM™ I Reduced Serum Medium

Brand:  Gibco™ 31985054

214.01 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


276.30 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 12087549

  • £321.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications. The complete formulation is confidential. For more information, please contact Technical Services.

Using Opti-MEM™ I Medium
Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

cGMP manufacturing and quality system
Opti-MEM™ I is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Opti-MEM™ I product made in our Grand Island facility. This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 X
Reduced-Serum Medium
Mammalian cell culture, transfection
ISO 13485 standards, FDA
2 to 8°C
Liquid
Glutamine, Phenol Red, Sodium Pyruvate
10 x 500 mL
7.0 to 7.4
Opti-Mem™ Reduced-Serum Medium, 10 x 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With L-Glutamine, Phenol Red, Sodium Pyruvate, 18 Months Shelf Life, 280 to 320mOsm/kg Osmolality, For Mammalian Cell Culture, Transfection
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Opti-MEM™ I Reduced Serum Medium > 10 x 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.