missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Opti-MEM™ I Reduced Serum Medium

Opti-MEM™ I Reduced Serum Medium
£32.46 - £321.00
Specifications
Concentration | 1 X |
---|---|
Form | Liquid |
Product Type | Reduced-Serum Medium |
With Additives | Glutamine, Phenol Red, Sodium Pyruvate |
For Use With (Application) | Mammalian cell culture, transfection |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11524456
![]() |
Gibco™
31985047 |
500 mL |
£32.46
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12087549
![]() |
Gibco™
31985054 |
10 x 500 mL |
£321.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications. The complete formulation is confidential. For more information, please contact Technical Services.Using Opti-MEM™ I Medium
Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
cGMP manufacturing and quality system
Opti-MEM™ I is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Opti-MEM™ I product made in our Grand Island facility. This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standards.
Specifications
1 X | |
Reduced-Serum Medium | |
Mammalian cell culture, transfection | |
7.0 to 7.4 |
Liquid | |
Glutamine, Phenol Red, Sodium Pyruvate | |
ISO 13485 standards, FDA | |
2 to 8°C |
Safety and Handling
missing translation for 'shelfLife' : 18 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Opti-MEM™ I Reduced Serum Medium