missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ 1 Capillary Column for GC

Brand:  Macherey-Nagel™ 726081.25

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10012851

  • £510.00 / Each
Estimated Shipment: 10-09-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.25 mm
GC Capillary Column
Optima™ 1
340°C to 360°C
25 m
G1, G2, G38
0.4 mm
Separation of Components According to Boiling Points Analysis
0.5 μm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers