Learn More
Macherey-Nagel™ Optima™ 1 Capillary Column for GC
£235.00 - £880.00
Specifications
Diameter (Metric) | 0.4 mm |
---|---|
Stationary Phase | Optima™ 1 |
USP Type | G1, G2, G38 |
Type | GC Capillary Column |
Temperature Range | 340°C to 360°C |
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
10798422
|
Macherey-Nagel™
726025.20 |
0.1 mm | 0.4 μm | 20 m |
£469.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10442042
|
Macherey-Nagel™
726038.15 |
0.25 mm | 0.1 μm | 15 m |
£315.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10421852
|
Macherey-Nagel™
726024.20 |
0.1 mm | 0.1 μm | 20 m |
£469.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10398441
|
Macherey-Nagel™
726038.10 |
0.25 mm | 0.1 μm | 10 m |
£235.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10516331
|
Macherey-Nagel™
726050.10 |
0.25 mm | 0.25 μm | 10 m |
£235.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10239551
|
Macherey-Nagel™
726024.10 |
0.1 mm | 0.1 μm | 10 m |
£284.00
Pack of 10 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10516681
|
Macherey-Nagel™
726050.15 |
0.25 mm | 0.25 μm | 15 m |
£315.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10368591
|
Macherey-Nagel™
726081.10 |
0.25 mm | 0.5 μm | 10 m |
£235.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10679761
|
Macherey-Nagel™
726038.25 |
0.25 mm | 0.1 μm | 25 m |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10154461
|
Macherey-Nagel™
726050.25 |
0.25 mm | 0.25 μm | 25 m |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10012851
|
Macherey-Nagel™
726081.25 |
0.25 mm | 0.5 μm | 25 m |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10596151
|
Macherey-Nagel™
726038.30 |
0.25 mm | 0.1 μm | 30 m |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10145181
|
Macherey-Nagel™
726081.30 |
0.25 mm | 0.5 μm | 30 m |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10114511
|
Macherey-Nagel™
726050.30 |
0.25 mm | 0.25 μm | 30 m |
£510.00
Pack of 30 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
10490122
|
Macherey-Nagel™
726050.50 |
0.25 mm | 0.25 μm | 50 m |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10556531
|
Macherey-Nagel™
726802.25 |
0.25 mm | 1 μm | 25 m |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10256851
|
Macherey-Nagel™
726802.50 |
0.25 mm | 1 μm | 50 m |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10535302
|
Macherey-Nagel™
726802.30 |
0.25 mm | 1 μm | 30 m |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10347971
|
Macherey-Nagel™
726081.50 |
0.25 mm | 0.5 μm | 50 m |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10003001
|
Macherey-Nagel™
726038.60 |
0.25 mm | 0.1 μm | 60 m |
£880.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10296651
|
Macherey-Nagel™
726050.60 |
0.25 mm | 0.25 μm | 60 m |
£880.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10022851
|
Macherey-Nagel™
726081.60 |
0.25 mm | 0.5 μm | 60 m |
£880.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10726132
|
Macherey-Nagel™
726802.60 |
0.25 mm | 1 μm | 60 m |
£880.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10164561
|
Macherey-Nagel™
726837.50 |
0.2 mm | 0.35 μm | 50 m |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10206811
|
Macherey-Nagel™
726834.12 |
0.2 mm | 0.2 μm | 12 m |
£265.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10595541
|
Macherey-Nagel™
726834.25 |
0.2 mm | 0.2 μm | 25 m |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10154561
|
Macherey-Nagel™
726837.12 |
0.2 mm | 0.35 μm | 12 m |
£265.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10525391
|
Macherey-Nagel™
726834.50 |
0.2 mm | 0.2 μm | 50 m |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10716332
|
Macherey-Nagel™
726837.25 |
0.2 mm | 0.35 μm | 25 m |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10032911
|
Macherey-Nagel™
726839.50 |
0.2 mm | 0.5 μm | 50 m |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
Specifications
0.4 mm | |
G1, G2, G38 | |
340°C to 360°C |
Optima™ 1 | |
GC Capillary Column | |
1 Each |