Learn More
Macherey-Nagel™ Optima™ 1 Capillary Column for GC
Brand:  Macherey-Nagel™ 726802.50
 View more versions of this product
Product Code. 10256851
  • £854.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
0.25 mm
G1, G2, G38
340°C to 360°C
50 m
GC Capillary Column
Optima™ 1
Separation of Components According to Boiling Points Analysis
1 μm
1 Each