Learn More
Thermo Scientific™ Optochin Disc
Thermo Scientific™ Oxoid Optochin Disc is for the differentiation of Streptococcus pneumoniae.
Brand: Thermo Scientific™ DD0001T
10.81 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Thermo Scientific™ Oxoid Optochin Disc is for the differentiation of Streptococcus pneumoniae.
Ordering Alerts:
5μg. 50 Discs/Cartridge. 1 Cart/Pk.
Specifications
Optochin (Ethylhydrocuprein Hydrochloride) | |
5µg | |
Microbial Differentiation | |
1 Cartridge/Pk. |
Streptococcus pneumoniae | |
Optochin Disc | |
Paper | |
50 Discs/Cartridge |