missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Optochin Disc

Thermo Scientific™ Oxoid Optochin Disc is for the differentiation of Streptococcus pneumoniae.

£19.80 - £78.45

Specifications

Species Streptococcus pneumoniae
Antimicrobial Agents Optochin (Ethylhydrocuprein Hydrochloride)
Concentration 5µg
Product Type Optochin Disc
Test Type Microbial Differentiation
View More Specs

Products 2
Product Code Brand Concentration Packaging Type Quantity Price Quantity & Availability  
Product Code Brand Concentration Packaging Type Quantity Price Quantity & Availability  
11429858
View Documents
Thermo Scientific™
DD0001B
5µg 5 Cartridges/Pk. 50 Discs/Cartridge
£78.45
Pack of 250
In Stock
to see stock.
Add to basket
 
11903842
View Documents
Thermo Scientific™
DD0001T
5µg 1 Cartridge/Pk. 50 Discs/Cartridge
£19.80
Pack of 50
In Stock
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ Oxoid Optochin Disc is for the differentiation of Streptococcus pneumoniae.

Differentiate Streptococcus pneumoniae from other streptococci using Thermo Scientific™ Oxoid™ Optochin Disc. Optochin Discs are paper discs ready impregnated with Optochin (ethylhydrocuprein hydrochloride) which provide a convenient and reliable alternative to the bile-solubility test. Pneumococciare are sensitive to Optochin so that a culture shows a zone of inhibition around the impregnated disc, while streptococci either grow right up to the edge of the disc or, occasionally, show a very small zone of inhibition.

Ordering Alerts:

5μg. 50 Discs/Cartridge. 1 Cart/Pk.

Specifications

Specifications

Streptococcus pneumoniae
5µg
Microbial Differentiation
50 Discs/Cartridge
Optochin (Ethylhydrocuprein Hydrochloride)
Optochin Disc
Paper
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.