Learn More
Quickfit™ 27BU Organic Chemistry Kit
Ideal organic chemistry kit for use in schools, colleges, and universities. Quickfit™ 27BU Organic Chemistry Kit is perfect for introducing students to the principal techniques of organic chemistry.
Brand:  Quickfit™ 27BU
333.02 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12433461
  • £339.00 / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ideal organic chemistry kit for use in schools, colleges, and universities. Quickfit™ 27BU Organic Chemistry Kit is perfect for introducing students to the principal techniques of organic chemistry.
  • The most popular Quickfit™ set for schools, colleges and universities
  • Comprises nine basic items of Quickfit™ glassware all with size 14/23 ground glass joints for compatibility
  • Allows the construction of at least 8 assemblies including steam distillation, reflux with addition and preparation
  • Scale based on starting material quantities up to 30g
  • Housed in a robust cardboard box with foam rubber liner to provide safe storage and transit
  • Supplied with a CD-ROM which includes practical details of some typical assemblies
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Organic Chemistry Kit
Pear shaped flask, Stillhead, Liebig condenser, Screwcap adapter, Receiver adapter, Air leak/steam inlet tube, Dropping funnel with GP Rotoflo tap, Stopper, Thermometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ 27BU Organic Chemistry Kit > Product Type: Organic Chemistry Kit