Learn More
Thermo Scientific™ Orion™ ROSS™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode
Measure flat, soft, moist, semi-solid or small volume samples with epoxy body flat surface pH electrodes.
Brand: Thermo Scientific™ 8135SC
Description
The 8135BN ROSS Combination Flat Surface pH Electrode measures pH of soft, moist surfaces — both solid and semi-solid —such as agar gel plates, meats, bread dough and similar samples. With drift less than 0.002 pH per day and minimized restandardization, this electrode works in extreme cases of samples varying from one another by 50°C or more.
- Material: Epoxy body
- pH range: 0 to 14
- Temperature range: 0° to 100°C
- Internal reference: ROSS
- Junction: Glass fiber
- Dimensions: 120 × 12mm
- pH precision: 0.01
- Fill solution: Orion 810007
- Cap diameter: 16mm
- Small samples are available in micro sample dish
- Flat surface tip
Includes: One 60 ml bottle of ROSS fill Solution, 810007 and spout cap

Specifications
With screw cap connector | |
Glass Fiber | |
0 to 14 | |
Combination Flat Surface pH Electrode, screw cap connector | |
3M KCl | |
ROSS | |
Short-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode. | |
0.47 in. | |
16 mm |
Screw Cap | |
Epoxy Body | |
0°C to 100°C | |
<0.002pH/day | |
Standard pH Meter, with separate cable with screw cap connector | |
0.01 | |
32°F to 212°F | |
12 mm |

Your input is important to us. Please complete this form to provide feedback related to the content on this product.