missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ ROSS™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode
Measure flat, soft, moist, semi-solid or small volume samples with epoxy body flat surface pH electrodes.
£295.81 - £405.29
Specifications
Junction | Glass Fiber |
---|---|
Material | Epoxy Body |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 100°C |
Drift | <0.002pH/day |
Product Code | Brand | Description | Price | Quantity | Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity | Availability | |||||
10468932
|
Thermo Scientific™
8135BN |
With BNC connector | With BNC connector |
£295.81
Each |
In Stock
Log in to see stock. |
|||||
10448722
|
Thermo Scientific™
8135SC |
With screw cap connector | With screw cap connector | N/A | ||||||
Description
Thermo Scientific™ Orion™ ROSS Flat Surface Electrodes have a uniquely designed, flat sensing bulb that allow for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates.
Orion™ ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
Solutions required
Electrode fill solution
810007 ROSS pH electrode fill solution, 5 x 60 mL bottles
Electrode storage solution
810001 ROSS pH electrode storage solution, 475 mL bottle
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
storage bottle
electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
810007 ROSS pH electrode fill solution 60 mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
Glass Fiber | |
0 to 14 | |
<0.002pH/day | |
ROSS | |
Short-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode. | |
0.47 in. | |
16 mm |
Epoxy Body | |
0°C to 100°C | |
3M KCl | |
0.01 | |
32°F to 212°F | |
12 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Orion™ ROSS™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode