missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ Sure-Flow™ Double Junction Epoxy Bodied Reference Electrode
Attain stable, drift-free reference measurements using a sure-flow electrode with its unique free-flowing liquid-to-liquid junction.
Brand: Thermo Scientific™ 900200
223.01 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
The easy-to-clean junction never clogs, simply press the cap and flush the junction area, then release the cap and the junction is reset. Even the most problematic, dirty or viscous samples can be easily measured without a clogged junction.
Thermo Scientific™ Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples.
Solutions required
Electrode fill solution
900002 Inner chamber fill solution for 900200, 5 x 60 mL
900003 Outer chamber fill solution for 900200, 5 x 60 mL
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
storage bottle
electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
900002 Double junction reference inner fill solution 60mL 1 bottle
900003 Double junction reference outer fill solution 60mL 1 bottle
Warranty period
12 months from date of purchase

Specifications
900200 Sure-Flow Reference Half-Cell Electrode | |
Sure-Flow | |
0 to 15 | |
Reference Half-Cell Electrode | |
Ag/AgCl Double Junction | |
1 m | |
16 mm |
Pin-tip | |
Epoxy Body | |
0°C to 100°C | |
variety of filling solutions | |
110 mm | |
12 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
PS, PT, PY Antibody Sampler Pack
Version: 12/08
File size: 33.9 kB
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
24Hours
13Minutes
8Seconds
32
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Orion™ Sure-Flow™ Double Junction Epoxy Bodied Reference Electrode > Internal reference: Ag/AgCl double junction
Spot an opportunity for improvement?Share a Content Correction