missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ 927006MD Glass ATC Probe

Automatically correct pH measurements for variation in electrode slope due to temperature change with this glass ATC probe.

Brand:  Thermo Scientific™ 927006


 View more versions of this product

Product Code. 10010263

  • £353.45 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Automatically correct pH measurements for variation in electrode slope due to temperature change with the Thermo Scientific™ Orion™ 927006MD Glass ATC Probe. Available with three different connector options.

Note:
Three different models are available to accommodate different Orion meters. MiniDIN version is compatible with older meters with use of an adapter cable.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

3.5 mm Phono Jack connector (ATC)
Orion Star Series Meters
0°C to 100°C
0 to 14
8 mm
120 mm
3.5 mm Phono Jack
±1°C
Glass
ATC Probe, Glass
0.3 in.
4.7 in.
32°F to 212°F
Store in air
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ATC PROBE ORION(TM) PERPHECT

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-3(2H)-isothiazolone

SDS
Documents

Documents

Certificates
Special Offers

Special Offers