missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits

Product Code. 11762218
Click to view available options
Type:
Alkalinity
Aluminum
Ammonia
Bromine Reagent Tablet
Chloride
Chlorine
Chlorine Dioxide
Chlorine, Sodium Chlorate
Copper
Cyanuric Acid
Hydrazine
Iron
Manganese
Metallphthalein Method
Nitrate
Nitrite
Ozone
Phosphate
Silica
Sulfate
Sulfide
Zinc
pH
Range:
0.01 to 0.25 mg/L
0.01 to 0.3 mg/L Al
0.01 to 0.5 mg/L N
0.01 to 6 mg/L Cl2
0.02 to 1 mg/L Fe
0.02 to 1 mg/L N
0.02 to 1 mg/L Zn
0.02 to 11 mg/L ClO2
0.04 to 0.5 mg/L S
0.05 to 0.5 mg/L N2H4
0.05 to 0.5 mg/L O3
0.05 to 4 mg/L PO4
0.05 to 4 mg/L SiO2
0.05 to 5 mg/L Cu
0.1 to 13.5 mg/L
0.2 to 4 mg/L Mn
0.5 to 25 mg/L Cl-
1 to 40 mg/L
1 to 80 mg/L PO4
2 to 50 mg/L
5 to 100 mg/L SO4
5 to 200 mg/L
5 to 200 mg/L CaCO3
5 to 90 mg/L
6.5 to 8.4 pH
Quantity:
100 DPD No 1 Tablets
100 DPD No 4 Tablets
100 Tablets
100 Tests
30 Tests
50 Tests
50 Tests Each (free and total)
Unit Size:
Pack of 100
Pack of 30
Pack of 50
This item is not returnable. View return policy

Product Code. 11762218

Brand: Thermo Scientific™ AC2016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.

Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100
TRUSTED_SUSTAINABILITY

Specifications

Type Sulfide
Test Reagent Formats Tablet
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
Description AQUAfast sulfide reagent tablets, 100 tests
Includes Set of 2: 100 Sulfide No 1 Tablets, 100 Sulfide No 2
Method DPD/Catalyst
For Use With (Application) Colorimetric water and wastewater testing equipment
Recommended Accessories AQUAfast 24mm round vials (Cat. No. AC2V24)
Range 0.04 to 0.5 mg/L S
Quantity 100 Tests
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308-IF exposed or concerned:.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : sodium bisulphate, boric acid, adipic acid, potassium chromate, potassium dichromate

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.