missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Basic Electrodes

Perform basic pH measurements quickly and reliably with Thermo Scientific™ Orion™ Basic Electrodes.

Brand:  Thermo Scientific™ 9142BN

84.85 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 11742837

  • £99.82 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform basic pH measurements quickly and reliably with Thermo Scientific™ Orion™ Basic Electrodes, for use with Orion™ Russell meters.

  • 0-14pH range
  • 0° to 80°C temperature range for pH electrodes
  • 0° to 50°C temperature range for 9147BN
  • 9142BN, 9145BN and 9146BN can be used with any meter with BNC input
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Refillable combination pH electrode with glass body
Glass Body
0°C to 80°C
Russell meters
32°F to 176°F
BNC
0 to 14
Combination pH Electrode, glass body
Store in Orion pH electrode storage solution 910001
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.