missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Basic Electrodes

Perform basic pH measurements quickly and reliably with Thermo Scientific™ Orion™ Basic Electrodes.

£75.03 - £128.21

Specifications

pH Range 0 to 14
For Use With (Equipment) Russell meters
Storage Requirements Store in Orion pH electrode storage solution 910001
View More Specs

Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11742837
View Documents Promotion Details
Thermo Scientific™
9142BN
Refillable combination pH electrode with glass body
£99.82
Each
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
11752837
View Documents Promotion Details
Thermo Scientific™
9146BN
Low maintenance gel combination with epoxy body and fluted guard
£75.03
Each
In Stock
to see stock.
Add to basket
 
11762837
View Documents Promotion Details
Thermo Scientific™
9147BN
Low maintenance gel combination w/epoxy body, built-in ATC
£128.21
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Perform basic pH measurements quickly and reliably with Thermo Scientific™ Orion™ Basic Electrodes, for use with Orion™ Russell meters.

  • 0-14pH range
  • 0° to 80°C temperature range for pH electrodes
  • 0° to 50°C temperature range for 9147BN
  • 9142BN, 9145BN and 9146BN can be used with any meter with BNC input
Warranty: 6 months from date of purchase by Buyer or 12 months from date of shipment from Seller, whichever is earlier.

Specifications

Specifications

0 to 14
Russell meters
Store in Orion pH electrode storage solution 910001
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.