Learn More
Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes
Help ensure proper and optimal probe performance with the Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes.
Brand: Thermo Scientific™ 080160
Code : 59
Additional Details : Weight : 0.01000kg
Description
Help ensure proper and optimal probe performance with Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes. Adapters include stir funnel and electrode adapter sleeve for BOD bottle.
Warranty: Out of box.
Specifications
BOD Adapter | |
Orion Dissolved Oxygen Probe (lab or field) | |
BOD Adapter | |
Stir funnel and electrode adaptor sleeve |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.