Learn More
Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents.
Brand: Thermo Scientific™ 977010
Additional Details : CAS Number : 1310-73-2 Weight : 0.80000kg
Description
Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.
Warranty: Out of box
Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements. The ammonia pH-adjusting ISA (951211) and low-level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low-level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.
All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:
- Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
- Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
- Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
- Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
Specifications
10 ppm | |
Ion Selective Electrode | |
Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes |
Iodide Reagent for Residual Chlorine Electrode | |
Ion Selective Electrode Reagents | |
5 x 50 mL |
Safety and Handling
- Iodide Reagent for Residual Chlorine
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P235-Keep cool.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see supplemental instructions on the administration of antidotes on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : potassium iodide, sodium hydroxide
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.