Learn More
Thermo Scientific™ Orion Lab Star PH111 Bench pH Meters
Orion Lab Star PH111 benchtop pH meters measure pH or ORP with mV and temperature, feature a large backlit display for easy viewability, and offer small footprint to save bench space. The Orion Lab Star pH meter is ideal for simplified, budget-friendly lab testing and data recording.
£574.74 - £881.50
Specifications
Accuracy (mV) ±0.2 or ±0.05%, whichever is greater
Accuracy (Temperature) ±0.3°C/±0.5°F
Data Management Details Transfer single or all data points to printer or computer
Outputs Computer or printer (STARA-106)
Measurement Modes pH with mV and Temperature, ORP with mV and Temperature
View More Specs
Products 5
Product Code Brand Description Includes Price Quantity & Availability  
Product Code Brand Description Includes Price Quantity & Availability  
17731849
View Documents Promotion Details
Thermo Scientific™
LSTAR1110
Orion Lab Star PH111 bench pH/mV meter with stand Meter with electrode stand, PC cable, universal power adapter
£574.74
Each
In Stock
to see stock.
Add to basket
17741849
View Documents Promotion Details
Thermo Scientific™
LSTAR1115
Orion Lab Star PH111 bench pH/mV meter standard kit 9157BNMD refillable epoxy-body pH/ATC electrode 916099 pH 4/7/10 buffer, storage and cleaning solution 60 mL kit meter with electrode stand, PC cable, universal power adapter
£788.06
Each
17751849
View Documents Promotion Details
Thermo Scientific™
LSTAR1116
Orion Lab Star PH111 bench pH/mV meter easy-to-clean bio kit GD9156BNWP double junction refillable epoxy-body pH electrode 927007MD stainless steel ATC temperature probe 916099 pH 4/7/10 buffer, storage and cleaning solution 60 mL kit meter with electrode stand, PC cable, universal power adapter
£814.53
Each
17761849
View Documents Promotion Details
Thermo Scientific™
LSTAR1117
Orion Lab Star PH111 bench pH/mV meter low-maintenance bio kit GD9106BNWP double junction gel-filled epoxy-body pH electrode 927007MD stainless steel ATC temperature probe 916099 pH 4/7/10 buffer, storage and cleaning solution 60 mL kit meter with electrode stand, PC cable, universal power adapter N/A N/A N/A
17771849
View Documents Promotion Details
Thermo Scientific™
LSTAR1118
Orion Lab Star PH111 bench pH/mV meter difficult sample kit 9165 BNWP Sure-Flow refillable epoxy-body pH electrode 927007MD stainless steel ATC temperature probe 916099 pH 4/7/10 buffer, storage and cleaning solution 60 mL kit meter with electrode stand, PC cable, universal power adapter
£881.50
Each
Description
Show More
Specifications
Show More
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Orion Lab Star PH111 Bench pH Meters