missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Nitrate Electrodes

Measure nitrate in aqueous solutions simply, accurately and economically with these electrodes.

Brand:  Thermo Scientific™ 930702

Code : 59

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10709361

  • £122.19 / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

Measure nitrate in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Nitrate Electrodes.

9707BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. To be used with module 970701.
Nitrate Half-Cell Electrode 9307BNWP (comes with half cell module 930702) is to be used with the Orion 900200 sure flow double junction reference electrode.

  • Calibration standards catalog number: 920706 — 0.1M NaNO3; 920707 — 1000ppm N; 930707 — 100ppm N
  • Required ISA catalog number: 930711 or 930710/nitrate ISS
Specifications

Specifications

Replacement Module for Nitrate Half-Cell Electrode 9307BNWP; 1/pk.
Free-flowing Sure-Flow™ junction assures stable
Combination Ion Selective Electrode
drift-free potentials
ISE Meters
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers