missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Electrode Filling Solution

Use this pH electrode filling solution with pH electrodes and reference electrodes.

£90.52 - £135.61

Specifications

Type pH Electrode Filling Solution
Quantity 60 mL
View More Specs

Products 10
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11767787
View Documents Promotion Details
Thermo Scientific™
510011
Filling Solution for No Cal pH Electrodes
£135.61
Pack of 5
In Stock
to see stock.
Add to basket
 
11777787
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
610011
Filling Solution for pHuture pH Electrodes
£123.62
Pack of 5
Estimated Shipment: 04-12-2024
to see stock.
Add to basket
 
12083363
View accessory products missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
810007
Filling Solution for ROSS Ultra, ROSS, PerpHecT ROSS Electrodes
£99.82
Pack of 5
In Stock
to see stock.
Add to basket
 
12064502
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
900001
Filling Solution for Single Junction Sure-Flow Reference Electrodes
£99.82
Pack of 5
In Stock
to see stock.
Add to basket
 
12034502
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
900002
Double Junction Sure-Flow Reference Electrode, Inner Chamber
£102.56
300mL
In Stock
to see stock.
Add to basket
 
12044502
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
900003
Double Junction Sure-Flow Reference Electrode, Outer Chamber
£102.56
Pack of 5
In Stock
to see stock.
Add to basket
 
11797787
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
900004
Standard, Specialty, PerpHecT Ag/AgCl and Single Junction Refillable Green pH Electrodes Used in Low Ionic Strength Samples
£105.84
Pack of 5
In Stock
to see stock.
Add to basket
 
11707797
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
900011
Standard, Specialty, PerpHecT Ag/AgCl, Single Junction Refillable Green and Micro pH Electrodes, ORP Electrodes
£101.72
Pack of 5
In Stock
to see stock.
Edge
Add to basket
 
11717797
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
900014
Filling solution for TRIS Calomel, TRIS Calomel KNIpHE electrodes This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13499216
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
910008-WA
Filling Solution for Double Junction, Double Junction KNIpHE™ Electrodes
£101.54
Pack of 5
Estimated Shipment: 04-12-2024
to see stock.
Add to basket
 
Description

Description

Use Thermo Scientific™ Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes. Packaged as five 2 oz. (5 x 60mL) bottles.

  • Only use ROSS™ filling solution (810007) with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
  • When the single junction reference electrode (900100) or double junction reference electrode (900200) are used with Orion ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide

Warranty: Out of box.

Specifications

Specifications

pH Electrode Filling Solution
60 mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Reference Filling Solution

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P321-Specific treatment (see supplemental first aid instructions on this label).

Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,boric acid, iodine

SDS
Documents

Documents

Certificates

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.