Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes for Orion Series Meters
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes.
Brand: Thermo Scientific™ 8157BNUMD
Description
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes. They are ideal for any application where the use of metallic reference electrodes would contaminate the samples. ROSS Ultra pH/ATC electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements. They have great stability and drift and come with a two-year replacement warranty.
Unique internal element system replaces Ag/AgCl or Hg2Cl2 internals with platinum wire and redox-couple filling solution. Double-junction reference cells have porous ceramic or glass fiber outer junction. Design provides stable potential, independent of temperature, and stable readings even when samples vary by 50°C (90°F) or more. Minimizes drift, stirring effects, clogged junctions; no need for frequent standardization so samples are analyzed faster.
- Epoxy body for durability and ruggedness
- Built-in ATC probe
- General-purpose applications
- Standard size and standard bulb
- Glass fiber junction
- L × Dia.: 120 × 12mm
Specifications
- 0 to 14pH
- 0° to 100°C (32° to 212°F) range
- Slope, 92 to 102%; isopotential point, pH 7
- When standardized in 6.86pH buffer at 25°C (77°F), accuracy is ±0.03pH for the buffer from 0° to 100°C (32° to 212°F) (using ATC); response is stable to 0.01pH within 30 seconds at 25°C (77°F) and 75°C (167°F) and stable to ±0.002pH within 15 seconds when cycling between pH 6.86 and 4.01 buffers at 25°C (77°F)
- Max. cap dia. (top): 0.75in. (19mm); collar dia. 0.62in. (16mm); Cable length is 39.37in. (1m)
Specifications
Orion ROSS Ultra Refillable pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 1 meter cable | |
Glass Fiber | |
0 to 14 | |
pH/ATC Combination Electrode | |
Orion Versa Star, Star A and Dual Star Meters | |
7 | |
0.01 | |
2 Years | |
1 m |
BNC Waterproof, 8-pin MiniDIN | |
Epoxy Body | |
0°C to 100°C | |
ROSS electrode 3 M KCl fill solution (Cat. No. 810007) | |
ROSS | |
0.03 pH for buffers using automatic temperature compensation | |
TRIS, sulfides, or proteins | |
120 mm | |
12 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.