missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes for Orion Series Meters

Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes.

£320.96 - £415.88

Specifications

Junction Glass Fiber
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type pH/ATC Combination Electrode
View More Specs

Products 3
Product Code Brand Connector Types Length (Metric) Cable For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Connector Types Length (Metric) Cable For Use With (Equipment) Price Quantity & Availability  
10206993
View accessory products View Documents
Thermo Scientific™
8157BNUMD
BNC Waterproof, 8-pin MiniDIN 1 m Orion Versa Star, Star A and Dual Star Meters
£353.66
Each
In Stock
to see stock.
Edge
Add to basket
 
10430085
View Documents
Thermo Scientific™
8157UWMMD
BNC Waterproof, 8-pin MiniDIN 3 m Orion Versa Star, Star A and Dual Star Meters This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15532405
View accessory products View Documents Promotion Details
Thermo Scientific™
8157BNURCA
BNC Waterproof, RCA 1 m Mettler Toledo Five and Seven series pH meters
£320.96
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes. They are ideal for any application where the use of metallic reference electrodes would contaminate the samples. ROSS Ultra pH/ATC electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements. They have great stability and drift and come with a two-year replacement warranty.

Measure pH and Temperature with One Electrode

Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes. They are ideal for any application where the use of metallic reference electrodes would contaminate the samples. ROSS Ultra electrodes offer the best temperature performance in repeated, varying temperatures of all other electrodes. They have great stability and drift and come with a two-year replacement warranty.

Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.

Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.

Target Markets:

  • Pharmaceutical/Biopharmaceutical
  • Food Manufacturing
  • Academic/Government
  • Environmental


Applications:

  • Quality Control
  • Research 
  • General Purpose
Specifications

Specifications

Glass Fiber
0 to 14
pH/ATC Combination Electrode
ROSS
0.03 pH for buffers using automatic temperature compensation
2 Years
12 mm
Epoxy Body
0°C to 100°C
ROSS electrode 3 M KCl fill solution (Cat. No. 810007)
7
0.01
120 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.