Learn More
Thermo Scientific™ Orion Star™ A121 Portable pH Meter
BIOPHARMA
Take simple pH and temperature measurements with our portable pH meter with easy operation for basic field testing.
Brand:  Thermo Scientific™ STARA1210
 View more versions of this product
Product Code. 11601449
  • ÂŁ415.56 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Take simple, routine pH, mV and temperature measurements with the Thermo Scientific™ Orion Star™ A121 Portable pH Meter. This meter is an easy-to-use and budget-friendly instrument for basic pH analysis in the field. Perform up to a 3 point pH calibration with easy recall of calibration points and slope. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. This waterproof meter has an IP67-rated housing for rugged protection in outdoor environments.
Thermo Scientific Orion Star A121 Portable pH Meters are a simple, economical solution for your routine outdoor sample analysis. Featuring an emphasized measure key, selectable read modes and measurement stability indicator, our basic Orion Star A121 pH Meters are designed to deliver measurements you can trust at an affordable price.
  • Measure pH, mV or relative mV with temperature
  • Perform up to a 3 point pH calibration with automatic buffer recognition or manual buffer entry
  • Simplified, easy-to-read display shows main measurement with temperature, stability indicator and electrode condition icon
  • User-friendly operation with basic calibration text prompts
  • Easily recall calibration data for procedure checks
  • Non-volatile memory preserves 50 point internal data log and meter settings, even with loss of power
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
Videos
  • Thermo Scientific - Orion meters - Taking water analysis to the next level
    Antonia Finlayson introduces Thermo Scientific Orion Star A and VERSA STAR meters for water analysis spanning pH, ISE, conductivity and dissolved oxygen measurements.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Thermo Scientific Orion meters Taking water analysis to the next level
    • Thermo Scientific - Orion meters - Taking water analysis to the next level
      Antonia Finlayson introduces Thermo Scientific Orion Star A and VERSA STAR meters for water analysis spanning pH, ISE, conductivity and dissolved oxygen measurements.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Thermo Scientific Orion meters Taking water analysis to the next level
      SDS
      Documents Certificates
      View accessory products
      Special Offers
      Special Offers
      Product Content Correction
      Product Title
      Thermo Scientific™ Orion Star™ A121 Portable pH Meter > pH/mV/Temperature Portable Meter only