Learn More
Thermo Scientific™ Orion Star™ A211 Benchtop pH Meter

Reliably measure pH, mV, ORP and temperature with our benchtop pH meter for advanced lab analysis and printer or computer interfacing.
£745.88 - £983.21
Specifications
Accuracy (mV) | ±0.2 or ±0.05%, whichever is greater |
---|---|
Accuracy (pH) | ±0.002 |
Accuracy (Temperature) | ±0.1°C |
Data Management Details | Transfer single, range or all data points to printer or computer |
Outputs | USB, RS-232 |
Product Code | Brand | Description | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | |||||
11661449
![]() |
Thermo Scientific™
STARA2110 |
Orion Star A211 pH benchtop meter with stand | Star A211 pH meter, electrode stand, 100-240V universal power adapter, computer cable |
£745.88
Each |
In Stock
Log in to see stock. |
|||||
15661717
![]() |
Thermo Scientific™
STARA2114 |
Orion Star A211 pH benchtop meter difficult sample kit with ROSS Sure-Flow pH electrode and ATC probe | Star A211 pH meter, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD stainless steel ATC probe, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable |
£983.21
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16211432
![]() |
Thermo Scientific™
STARA2117 |
Orion Star A211 pH benchtop meter standard kit with general purpose pH/ATC electrode | Star A211 pH meter, 9157BNMD epoxy-body pH/ATC electrode, 910199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable |
£863.20
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||||
15671717
![]() |
Thermo Scientific™
STARA2118 |
Orion Star A211 pH benchtop meter separate sensor kit with ROSS Ultra pH electrode and ATC probe | Star A211 pH meter, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD stainless steel ATC probe, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable |
£932.35
Each |
In Stock
Log in to see stock. |
|||||
15681717
![]() |
Thermo Scientific™
STARA2119 |
Orion Star A211 pH benchtop meter food and beverage kit with Sure-Flow pH electrode and ATC probe | Star A211 pH meter, 9165BNWP Sure-Flow epoxy-body pH electrode, 927007MD stainless steel ATC probe, 910199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable |
£889.97
Each |
In Stock
Log in to see stock. |
|||||
11671449
![]() |
Thermo Scientific™
STARA2115 |
Orion Star A211 pH benchtop meter TRIS kit with TRIS-compatible ROSS Ultra pH/ATC electrode | Star A211 pH meter, 8302BNUMD ROSS Ultra glass-body pH/ATC electrode, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable |
£906.92
Each |
In Stock
Log in to see stock. |
|||||
12923257
![]() |
Thermo Scientific™
STARA2116 |
Orion Star A211 pH benchtop meter durable sensor TRIS kit with TRIS-compatible ROSS Ultra epoxy-body pH/ATC electrode | Star A211 pH meter, 8157BNUMD ROSS Ultra epoxy-body pH/ATC electrode, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable |
£915.40
Each |
In Stock
Log in to see stock. |
|||||
Description
Record accurate and reliable pH, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A211 Benchtop pH Meter. This meter is ideal for a wide range of applications and advanced pH analysis in the lab. Perform up to a 5 point pH calibration, log up to 2000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
Thermo Scientific Orion Star A211 Benchtop pH Meters are easy-to-use, dependable laboratory meters with advanced features and functions. Selectable read modes and stability indicator give you flexibility and control over your testing. All critical data is shown with your measurements including temperature, date and time, read type, measurement stability, active calibration information and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates.
Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view and stirrer keys that ensure easy operation. All setup menus are in one location with direct access using a single shortcut key so you can quickly access, evaluate and change measurement and instrument settings.
- Measure pH, mV, relative mV or ORP with temperature
- Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
- Calibration editing option allows individual points to be fixed without a full recalibration
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe (catalog number 096019) to uniformly mix solutions
- Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
- Calibration log stores 10 most recent calibrations
- Non-volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with universal power adapter or optional AA batteries
- IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
- Meters include CE, TUV 3-in-1 and FCC Class A certifications
- 3-year replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
±0.2 or ±0.05%, whichever is greater | |
±0.1°C | |
USB, RS-232 | |
pH with calibration editing option, relative mV, ORP and temperature | |
0.001, 0.01, 0.1 | |
0.1mV | |
-5°C to +105°C | |
0.1°C, 0.1°F | |
1 point temperature offset calibration | |
Yes, selectable | |
2000 with Date and Time Stamp | |
High/low limit alarm, calibration due alarm | |
CE, TUV 3-1, FCC Class A | |
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) | |
2 lb. | |
10 per channel with password protection | |
Universal AC adapter (included) or 4 AA batteries (sold separately) | |
±2000.0mV | |
7.000pH | |
41°F to 113°F (ambient) | |
1 | |
800 hr. |
±0.002 | |
Transfer single, range or all data points to printer or computer | |
pH, mV, Relative mV, ORP with Temperature | |
-2 to 20 | |
±2000.0mV | |
1 to 5 | |
22.0°F to 221.0°F | |
Manual or automatic with ATC temperature probe | |
LCD, Graphic with Backlight | |
Non-volatile memory preserves data log, calibration log and meter settings | |
Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode | |
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) | |
IP-54 | |
0.9 kg | |
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys | |
Orion Star A211 pH/mV/RmV/ORP/Temperature Benchtop Meter | |
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean Available with Software Update | |
5 to 85%, non-condensing | |
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting | |
5°C to 45°C (ambient) | |
4 x AA (sold separately) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.