Learn More
Thermo Scientific™ Orion Star™ A214 pH/ISE Benchtop Meter
Reliably measure pH, ion concentration, mV, ORP and temperature with our pH/ISE benchtop meter for advanced lab analysis and interfacing.
Brand: Thermo Scientific™ STARA2145
1455.09 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : 59
Additional Details : Weight : 6.50000kg
Description
Record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A214 pH and ISE Benchtop Meter. This meter is ideal for a wide range of applications and advanced pH or ion analysis in the lab. Perform up to 5 point pH and ISE calibrations, log up to 2000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
Thermo Scientific Orion Star A214 pH/ISE benchtop meters are easy to use, dependable laboratory meters with advanced features and functions. Selectable read modes and stability indicator give you flexibility and control over your testing. All critical data is shown with your measurements including temperature, date and time, read type, measurement stability, active calibration information and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates.
Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view and stirrer keys that ensure easy operation. All setup menus are in one location with direct access using a single shortcut key so you can quickly access, evaluate and change measurement and instrument settings.
- Measure pH, ion concentration, mV, relative mV or ORP with temperature
- Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
- Perform up to a 5 point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb or none
- Calibration editing option allows individual points to be fixed without a full recalibration
- Advanced ISE features include non-linear selectable auto-blank and low range stability
- Select from a wide variety of Orion ion selective electrodes including electrodes that measure ammonia, ammonium, bromide, cadmium, calcium, carbon dioxide, chloride, chlorine, cupric, cyanide, fluoride, fluoroborate, iodide, lead, nitrate, potassium, silver, sodium, sulfate, surfactant and thiocyanate concentrations
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe (catalog number 096019) to uniformly mix solutions
- Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
- Calibration log stores 10 most recent calibrations
- Non-volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with universal power adapter or optional AA batteries
- IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
- Meters include CE, TUV 3-in-1 and FCC Class A certifications
- 3-year replacement warranty
Specifications
USB, RS232 | |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.002 | |
High/low limit alarm, calibration due alarm | |
800 hr. | |
pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ | |
1 to 5 | |
CE, TUV 3-1, FCC Class A | |
2000 with date and time stamp | |
1 | |
IP 54 | |
±2000.0 mV | |
0.0001 minimum, 1 to 3 significant digits (user selectable) | |
-5.0°C to +105.0°C | |
BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) | |
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys | |
Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m | |
10 per channel with password protection | |
Universal AC adapter (included) or 4 AA batteries (sold separately) | |
0.0001 to 19900 with selectable units of ppm, Molar, mg/L, %, ppb or none | |
±2000.0 mV | |
0.1mV | |
±0.1°C | |
5°C to 45°C (ambient) | |
22.0°F to 221.0° F | |
Manual or automatic with ATC temperature probe | |
24cm x 18cm x 11cm (9.5 in. x 7.1 in. x 4.3 in.) | |
0.9 kg |
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) | |
±0.2 mV or ±0.05% of reading whichever is greater | |
Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit | |
Yes, selectable | |
4 AA (sold separately) | |
1 point relative mV (RmV) mode, 1 point ORP mode | |
1 to 5 | |
Transfer single, range or all data points to printer or computer | |
Orion Star A214 pH/ISE benchtop meter kit with ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe, solutions and stand | |
LCD Graphic, Backlit | |
pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature | |
-2.000 to 20.000 | |
0.1/0.01/0.001 | |
Star A214 pH/ISE Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 096019 Stirrer Probe; 810199 ROSS pH Buffer Kit | |
7.000pH | |
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update | |
Non-volatile memory preserves data log, calibration log and meter settings | |
Orion Star A214 pH/ISE/mV/RmV/ORP/Temperature Benchtop Meter | |
pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8102BNUWP ROSS Ultra pH electrode, 927007MD Orion ATC probe, 096019 Orion Star stirrer probe) | |
±2000.0 mV | |
5 to 85 %, non-condensing | |
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting | |
41°F to 113°F (ambient) | |
1 point temperature offset calibration | |
0.1°C, 0.1°F | |
3 Year Meter | |
2 lb. |