Learn More
Thermo Scientific™ Orion Star™ A216 pH/Dissolved Oxygen Benchtop Multiparameter Meter
Reliably measure pH, dissolved oxygen and temperature with our pH/DO/RDO benchtop multiparameter meter for advanced lab analysis and interfacing.
£1333.76 - £2168.43
Specifications
Outputs | USB, RS232 |
---|---|
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) |
Accuracy (Barometer) | ±6mmHg |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
11621469
|
Thermo Scientific™
STARA2160 |
Orion Star A216 pH/RDO/DO benchtop meter with stand | Star A216 pH/RDO/DO meter, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
£1333.76
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
11611469
|
Thermo Scientific™
STARA2165 |
Orion Star A216 pH/RDO/DO benchtop meter kit with ROSS Ultra Triode epoxy-body pH/ATC electrode, polarographic DO sensor, solutions and stand | Star A216 pH/RDO/DO benchtop meter kit with ROSS Ultra Triode epoxy-body pH/ATC electrode, polarographic DO sensor, solutions and stand |
£2168.43
Each |
In Stock
Log in to see stock. |
||||
Description
Record accurate and reliable pH, mV, ORP, dissolved oxygen and temperature measurements with the Thermo Scientific™ Orion Star™ A216 pH/Dissolved Oxygen Benchtop Multiparameter Meter. This meter is ideal for a wide range of applications, including advanced pH and dissolved oxygen analysis in the lab. Use the two channels to measure pH and dissolved oxygen simultaneously or view each channel separately. Use an RDO optical or polarographic DO probe for ultimate flexibility in sensors. Log up to 2000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer.
Thermo Scientific Orion Star A216 pH/RDO/DO Benchtop Meters are easy-to-use, dependable laboratory meters with advanced features and functions. Selectable read modes and stability indicator give you flexibility and control over your testing. All critical data is shown with your measurements including temperature, date and time, read type, measurement stability, active calibration information and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates.
Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view and stirrer keys that ensure easy operation. All setup menus are in one location with direct access using a single shortcut key so you can quickly access, evaluate and change measurement and instrument settings.
- Combined convenience with the benefits of a pH meter and dissolved oxygen meter in one easy-to-use system
- Measure pH, mV, relative mV or ORP with temperature on channel one and measure dissolved oxygen in percent saturation or mg/L with temperature using RDO optical DO probes or polarographic DO probes on channel two
- Easily view individual measurements on a full screen display and quickly toggle to a split screen display to view multiple measurements simultaneously
- Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values and calibration editing option that allows individual points to be fixed without a full recalibration
- Calibrate dissolved oxygen using water-saturated air, air-saturated water, custom value (Winkler titration) and zero point options
- Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe (catalog number 096019) to uniformly mix solutions
- Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
- Calibration log stores 10 most recent calibrations
- Non-volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with universal power adapter or optional AA batteries
- IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
- Meters include CE, TUV 3-in-1 and FCC Class A certifications
- 3-year replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
USB, RS232 | |
±6mmHg | |
±0.002 | |
±0.1mg/L up to 8mg/L, ±0.2mg/L from 8 to 20mg/L, ±10% of reading up to 50mg/L | |
Yes, selectable | |
800 hr. | |
pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature | |
1 point relative mV (RmV) mode, 1 point ORP mode | |
CE, TUV 3-1, FCC Class A | |
2000 with date and time stamp | |
LCD Graphic, Backlit | |
pH, mV, Relative mV, ORP with Temperature, Dissolved Oxygen (% Saturation), Dissolved Oxygen (mg/L with Temperature) | |
±2000.0 mV | |
0.1/0.01/0.001 | |
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe) | |
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys | |
Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m | |
10 per channel with password protection | |
0.0 to 600.0 % saturation | |
0.1, 1 % saturation | |
±2 % saturation or ±2% of reading whichever is greater | |
Universal AC adapter (included) or 4 AA batteries (sold separately) | |
1mmHg | |
0.00 to 90.00mg/L | |
±2000.0 mV | |
0.1mV | |
0.01, 0.1mg/L | |
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting | |
41°F to 113°F (ambient) | |
1 point temperature offset calibration | |
0.1°C, 0.1°F | |
3 Year Meter | |
2 lb. |
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) | |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.2mg/L or ±2% of reading, whichever is greater | |
High/low limit alarm, calibration due alarm | |
400 to 850mmHg, automatic using built-in barometer or manual entry | |
4 AA (sold separately) | |
Water-saturated air, air-saturated water, manual (Winkler titration), zero point | |
1 to 5 | |
Transfer single, range or all data points to printer or computer | |
2 | |
IP 54 | |
0.0 to 600.0 % saturation | |
-2.000 to 20.000 | |
-5.0°C to +105.0°C | |
7.000pH | |
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update | |
Non-volatile memory preserves data log, calibration log and meter settings | |
Orion Star A216 pH/mV/RmV/ORP/RDO Optical/Dissolved Oxygen/Temperature Benchtop Meter | |
±2 % saturation ≤ 200 % saturation, ±10 % saturation > 200 % saturation | |
0.0 to 500.0 % saturation | |
0.1, 1 % saturation | |
400 to 850mmHg | |
±2000.0 mV | |
0.00 to 50.00mg/L | |
5 to 85 %, non-condensing | |
0.01, 0.1mg/L | |
0.0 to 45.0 ppt, automatic correction using manual entry of sample salinity | |
±0.1°C | |
5°C to 45°C (ambient) | |
22.0°F to 221.0° F | |
Manual or automatic with ATC temperature probe | |
24cm x 18cm x 11cm (9.5 in. x 7.1 in. x 4.3 in.) | |
0.9 kg |