Learn More
Thermo Scientific™ Orion Star™ A221 Portable pH Meter
Reliably measure pH, mV, ORP and temperature with our portable pH meter for advanced field analysis and printer or computer interfacing.
£494.99 - £1118.83
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
11631469
|
Thermo Scientific™
STARA2210 |
Orion Star A221 pH portable meter | Star A221 pH meter, 4 AA batteries, computer cable |
£494.99
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15505199
|
Thermo Scientific™
STARA2216 |
Orion Star A221 pH portable meter standard kit with general purpose low-maintenance pH/ATC electrode | Star A221 pH meter, 9107BNMD gel-filled epoxy-body pH/ATC electrode, pH 4/7/10 buffer and rinse pouches, storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable |
£777.24
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15612057
|
Thermo Scientific™
STARA2217 |
Orion Star A221 pH portable meter ORP kit with low-maintenance ORP/ATC electrode | Star A221 pH meter, 9179BNMD gel-filled ORP/ATC electrode, ORP standard, rinse pouches, storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable |
£833.19
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15622057
|
Thermo Scientific™
STARA2218 |
Orion Star A221 pH portable meter food kit with ROSS Spear-Tip pH electrode and ATC probe | Star A221 pH meter, 8163BNWP ROSS spear-tip pH electrode, 927007MD stainless steel ATC probe, pH 4/7/10 buffer and rinse pouches, ROSS storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable |
£1118.83
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11641469
|
Thermo Scientific™
STARA2215 |
Orion Star A221 pH portable meter kit with ROSS Ultra Triode low maintenance gel pH/ATC electrode, solutions, meter armor and field case | Star A221 pH meter, 8107UWMMD ROSS Ultra gel-filled epoxy-body pH/ATC electrode with 3m cable, pH 4/7/10 buffer and rinse pouches, ROSS storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable |
£845.90
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Record accurate and reliable pH, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A221 Portable pH Meter. This meter is ideal for a wide range of applications and advanced pH analysis in the field. Perform up to a 5 point pH calibration, log up to 1000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Waterproof meter ensures durable, rugged performance in outdoor environments with an IP67-rated housing.
- Measure pH, mV, relative mV or ORP with temperature
- Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading and Continuous shows changing readings with hold option
- Data log collects and date/time stamps up to 1000 measurement sets
- Non-volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with AA batteries or optional universal power adapter
- IP67-rated housing is waterproof for rugged field operation
- Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
- Meters offer CE, TUV 3-in-1 and FCC Class A certifications
- 3-year replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.