missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion Star™ A321 pH Portable Meter
BIOPHARMA

Accurately monitor pH, mV, ORP and temperature with our waterproof, portable pH meter for high performance in challenging field locations.

Brand:  Thermo Scientific™ STARA3215

846.72 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 11601479

  • £1059.49 / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Get high accuracy and premium performance everywhere you need it with the Thermo Scientific™ Orion Star™ A321 pH Portable Meter. This meter is designed for a wide range of pH, mV, ORP and temperature testing and field applications. Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Perform up to a 5 point pH calibration, log up to 5000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software.

Take your measurements at the source with our premium line of portable meters. The Thermo Scientific Orion Star A321 pH Portable Meter offers an information-rich screen and user-friendly operation. This rugged, waterproof portable meter is ready to tackle the elements with you. Orion Star A321 pH Portable Meters are designed to be easy-to-use, dependable field meters that offer advanced features and functions including a 5000 point date log with sample ID and user ID options, calibration and high/low limit alarms and password-protected measurement methods.
  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
  • Calibration log stores 10 most recent calibrations
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
±0.2 mV or ±0.05% of reading whichever is greater
High/low limit alarm, calibration due alarm
800 hr.
pH with calibration editing option, relative mV (RmV), ORP and temperature
CE, TUV 3-1, FCC Class A
5000 with date and time stamp
1
IP 67
±2000.0 mV
0.1/0.01/0.001
Star A321 pH meter, 8107UWMMD ROSS Ultra gel-filled epoxy-body pH/ATC electrode with 3m cable, pH 4/7/10 buffer and rinse pouches, ROSS storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable
7.000pH
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Non-volatile memory preserves data log, calibration log and meter settings
Orion Star A321 pH/mV/RmV/ORP/Temperature Portable Meter
pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe (includes 8107UWMMD ROSS Ultra Triode pH/ATC electrode)
±2000.0 mV
0.1mV
±0.1°C
5°C to 45°C (ambient)
22.0°F to 221.0° F
Manual or automatic with ATC temperature probe
24cm x 10cm x 6.5cm (9.5 in. x 3.9 in. x 2.6 in.)
0.5 kg
USB, RS-232
±0.002
Yes, selectable
4 x AA
1 to 5
Transfer single, range or all data points to printer or computer
Orion Star A321 pH portable meter TRIS kit with ROSS Ultra low-maintenance pH/ATC electrode
LCD Graphic, Backlit
pH, mV, Relative mV, ORP with Temperature
-2 to 20
-5.0°C to +105.0°C
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m
10 per channel with password protection
4 AA batteries (included) or universal AC adapter (sold separately)
±2000.0 mV
5 to 85%, non-condensing
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
41°F to 113°F (ambient)
1 point temperature offset calibration
0.1°C, 0.1°F
3 Year Meter
1.2 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.