Learn More
Thermo Scientific™ Orion Star™ A325 pH/Conductivity Portable Multiparameter Meter
Accurately measure pH, conductivity and temperature with our pH/conductivity portable meter for high performance in challenging field tests.
£1023.98 - £1523.06
Specifications
Outputs | USB, RS232 |
---|---|
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003) |
Accuracy (Conductivity) | 0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
11671479
|
Thermo Scientific™
STARA3250 |
Orion Star A325 pH/conductivity portable meter | Star A325 pH/conductivity meter, 4 AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate |
£1023.98
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11681479
|
Thermo Scientific™
STARA3255 |
Orion Star A325 pH/conductivity portable meter kit with ROSS Ultra Triode low maintenance gel pH/ATC electrode, DuraProbe 4-cell conductivity sensor, solutions, meter armor and field case | Orion STAR A325 pH/conductivity portable meter; 8107UWMMD Orion ROSS Triode pH/ATC gel-filled electrode with 3m cable; 013010MD Orion DuraProbe conductivity cell with 3m cable; 910410 pH 4.01 buffer pouches, pack of 10; 910710 pH 7.00 buffer pouches, pack of 10; 911010 pH 10.01 buffer pouches, pack of 10; 01100710 conductivity standard pouches, 1413 uS; 911110 rinse solution, pack of 10; protective armor with electrode holder; hard carrying case and batteries |
£1523.06
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Get high accuracy and premium performance everywhere you need it with the Thermo Scientific™ Orion Star™ A325 pH/Conductivity Portable Multiparameter Meter. This meter is designed for a wide range of pH, mV, ORP, conductivity, total dissolved solids (TDS), salinity, resistivity and temperature testing and field applications. Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Use the two channels to measure pH and conductivity simultaneously or view each channel separately. Log up to 5000 data point sets with time/date stamp and transfer logs to a printer or computer.
- Combined convenience with the benefits of a pH meter and conductivity meter in one easy-to-use system
- Measure pH, mV, relative mV or ORP with temperature on channel one and measure conductivity, TDS, salinity or resistivity with temperature on channel two
- Easily view individual measurements on a full screen display and quickly toggle to a split screen display to view multiple measurements simultaneously
- Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
- Perform up to a 5 point conductivity calibration
- Calibration editing option allows individual points to be fixed without a full recalibration
- Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
- Use linear, non-linear, EP (USP) or off conductivity temperature compensation with 5°C, 10°C, 15°C, 20°C or 25°C reference temperatures
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
- Calibration log stores 10 most recent calibrations
- Non-volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with AA batteries or optional universal power adapter
- IP67-rated housing is waterproof for rugged field operation
- Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
- Meters offer CE, TUV 3-in-1 and FCC Class A certifications
- 3-year replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
USB, RS232 | |
0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS | |
±0.002 | |
0.5% of reading ±1 digit | |
High/low limit alarm, calibration due alarm | |
800 hr. | |
pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op | |
Up to 5 | |
0.001 to 199.9cm-1 | |
Transfer single, range or all data points to printer or computer | |
2 | |
IP 67 | |
0.001 μS/cm to 3000 mS/cm | |
-2.000 to 20.000 | |
0.06 to 80.00 psu, 0.05 to 42.00 ppt | |
0.1/0.01/0.001 | |
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe) | |
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys | |
Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m | |
10 per channel with password protection | |
4 AA batteries (included) or universal AC adapter (sold separately) | |
±2000.0 mV | |
0.001 μS minimum, auto ranging up to 4 significant digits | |
1 ohm or 0.1 meg-ohm, auto ranging | |
0.001 ppm minimum, auto ranging up to 4 significant digits | |
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting | |
±0.1°C | |
41°F to 113°F (ambient) | |
5, 10, 15, 20, 25°C | |
22.0°F to 221.0° F | |
Manual or automatic with ATC temperature probe | |
24cm x 10cm x 6.5cm (9.5 in. x 3.9 in. x 2.6 in.) | |
0.5 kg |
Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003) | |
±0.2 mV or ±0.05% of reading whichever is greater | |
0.5% of reading ±1 digit | |
0.5% of reading ±1 digit | |
Yes, selectable | |
4 AA (included) | |
1 point relative mV (RmV) mode, 1 point ORP mode | |
1 to 5 | |
CE, TUV 3-1, FCC Class A | |
5000 with date and time stamp | |
LCD Graphic, Backlit | |
pH, mV, Relative mV, TDS, ORP with Temperature, Salinity, Conductivity, Resistivity with Temperature | |
±2000.0 mV | |
2 ohm to 100.0 meg-ohm | |
0.001 ppm to 200.0 ppt | |
-5.0°C to +105.0°C | |
7.000pH | |
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update | |
Non-volatile memory preserves data log, calibration log and meter settings | |
Orion Star A325 pH/mV/RmV/ORP/Conductivity/TDS/Salinity/Resistivity/Temperature Portable Meter | |
±2000.0 mV | |
5 to 85 %, non-condensing | |
0.1mV | |
0.01 psu or 0.01 ppt minimum, auto ranging | |
Practical salinity (psu) or natural sea water (ppt) | |
Linear (0.02 to 9.99) | |
Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off | |
5°C to 45°C (ambient) | |
1 point temperature offset calibration | |
0.1°C, 0.1°F | |
3 Year Meter | |
1.2 lb. |