missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Total Alkalinity Test Kit

Save time by eliminating additional equipment set-up, titrations and calculations with this total alkalinity test kit that provides two-step direct measurement.

Brand:  Thermo Scientific™ 700010

 View more versions of this product

Product Code. 10140971

  • £232.33 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Save time by eliminating additional equipment set-up, titrations and calculations. The Thermo Scientific™ Orion™ Total Alkalinity Test Kit provides two-step direct measurement of total alkalinity and pH using a pH electrode.

Features:

  • Direct reading of total alkalinity from 0 to 200ppm as CaCO3, from the enclosed pH conversion chart
  • No interference from color or turbidity

Includes:

  • One 1 pint (475mL) bottle alkalinity reagent (700011)
  • One 1 pint (475mL) bottle alkalinity standard/control (700012)
  • Total alkalinity conversion chart
  • Application procedure No. 517, Total Alkalinity in Natural Waters

Note:

The following part number has been changed due to a recent system upgrade.

Old Part Number: 700011

New Part Number: 700011-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Data Safety Sheet (MSDS) files.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Total Alkalinity Test Kit
two-step direct measurement of total alkalinity and pH using a pH electrode; Saves time by eliminating additional equipment set-up, tedious titrations and calculations
475mL Alkalinity Reagent (700011), 475mL Alkalinity Standard/Control (700012), Total Alkalinity Conversion Chart and Application Procedure No. 517, Total Alkalinity in Natural Waters
475mL
Non Hazardous Media
Liquid
pH Electrode
Kit
Total Alkalinity Test Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.