missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oseltamivir phosphate, 98%, Thermo Scientific Chemicals
£50.65 - £118.59
Chemical Identifiers
CAS | 204255-11-8 |
---|---|
Molecular Formula | C16H28N2O4·H3PO4 |
Molecular Weight (g/mol) | 410.4 |
InChI Key | PGZUMBJQJWIWGJ-ONAKXNSWSA-N |
Synonym | oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1 |
PubChem CID | 78000 |
ChEBI | CHEBI:7799 |
IUPAC Name | ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid |
SMILES | CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15698715
|
Thermo Scientific Acros
461170010 |
1 g |
£50.65
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15608725
|
Thermo Scientific Acros
461170050 |
5 g |
£118.59
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
204255-11-8 | |
410.4 | |
oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1 | |
CHEBI:7799 | |
CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O |
C16H28N2O4·H3PO4 | |
PGZUMBJQJWIWGJ-ONAKXNSWSA-N | |
78000 | |
ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid |
Specifications
204255-11-8 | |
97.5% min. (HPLC) | |
Authentic | |
oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1 | |
CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O | |
410.4 | |
CHEBI:7799 | |
98% |
White to Off-White | |
C16H28N2O4·H3PO4 | |
Glass Bottle | |
PGZUMBJQJWIWGJ-ONAKXNSWSA-N | |
ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid | |
78000 | |
410.4 | |
Oseltamivir phosphate |
Safety and Handling
Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
RUO – Research Use Only