missing translation for 'onlineSavingsMsg'
Learn More

Osmium tetroxide, 4 wt.% solution in water, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 319010100

159.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 10145263

  • £209.04 / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100.0°C
20816-12-0, 7732-18-5
Solution
O4Os
MFCD00011150
15, 6990
Solubility in water: soluble
O=[Os](=O)(=O)=O
254.23
254.2
Glass bottle
1.0400g/mL
Osmium tetroxide
Yellow
10 mL
OsO4
01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
VUVGYHUDAICLFK-UHFFFAOYSA-N
tetraoxoosmium
30318
3.92 to 4.08%
1.04
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Osmium tetroxide, 4 wt% solution in water

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H310-Fatal in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only