missing translation for 'onlineSavingsMsg'
Learn More

Osmium tetroxide, 4 wt.% solution in water, Thermo Scientific Chemicals

£127.64 - £209.04

Specifications

Boiling Point 100.0°C
Chemical Name or Material Osmium tetroxide
CAS 20816-12-0, 7732-18-5
Color Yellow
Physical Form Solution
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10616881
View Documents
Thermo Scientific Acros
319010050
5 mL Glass bottle
£127.64
5mL
Please to purchase this item. Need a web account? Register with us today!
10145263
View Documents Promotion Details
Thermo Scientific Acros
319010100
10 mL Glass bottle
£209.04
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

100.0°C
20816-12-0, 7732-18-5
Solution
OsO4
01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
VUVGYHUDAICLFK-UHFFFAOYSA-N
tetraoxoosmium
30318
3.92 to 4.08%
1.04
Osmium tetroxide
Yellow
O4Os
MFCD00011150
15, 6990
Solubility in water: soluble
O=[Os](=O)(=O)=O
254.23
254.2
Glass bottle
1.0400g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Fatal in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air a

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 244-058-7

missing translation for 'rtecsNumber' : RN1140000

missing translation for 'tsca' : TSCA

Product Identifier
  • Osmium tetroxide, 4 wt% solution in water

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Osmium oxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only