Learn More
Osmium tetroxide, 4 wt.% solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 319010050
101.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
100.0°C | |
20816-12-0, 7732-18-5 | |
Solution | |
O4Os | |
MFCD00011150 | |
15, 6990 | |
Solubility in water: soluble | |
O=[Os](=O)(=O)=O | |
254.23 | |
254.2 | |
Glass bottle | |
1.0400g/mL |
Osmium tetroxide | |
Yellow | |
5 mL | |
OsO4 | |
01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236 | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
tetraoxoosmium | |
30318 | |
3.92 to 4.08% | |
1.04 |
Safety and Handling
- Osmium tetroxide, 4 wt% solution in water
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Osmium oxide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only