missing translation for 'onlineSavingsMsg'
Learn More

Osmium(VIII) oxide, 2% aq. soln., Thermo Scientific Chemicals

£73.97 - £124.62

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15442237
View Documents
Thermo Scientific Alfa Aesar
045384.9W
2 mL
£73.97
2mL
Please to purchase this item. Need a web account? Register with us today!
15452237
View Documents
Thermo Scientific Alfa Aesar
045384.AB
10 mL
£124.62
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Osmium(VIII) oxide is widely used as an oxidant in organic synthesis to oxidize alkenes to vicinal diols as well as in the sharpless oxyamination to give vicinal amino-alcohols. It is involved in the oxidative cleavage of alkenes (Lemieux-Johnson oxidation) in association with sodium periodate. It finds application in transmission electron microscopy (TEM) as a staining agent. It is involved in the promotion of oxidative cleavage of graphene layers in carbon nanotubes and as an intermediate in osmium ore refining. As a lipid stain, it is useful in scanning electron microscopy (SEM) as an alternative to sputter coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20816-12-0
254.23
VUVGYHUDAICLFK-UHFFFAOYSA-N
30318
O=[Os](=O)(=O)=O
O4Os
MFCD00011150
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
tetraoxoosmium

Specifications

20816-12-0
Liquid
MFCD00011150
14,6893
Soluble in alcohol,ether,chloroform,benzene,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride
O=[Os](=O)(=O)=O
254.23
254.1
Chlorine-like
Osmium (VIII) oxide
Colorless to Yellow
O4Os
UN3287
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
VUVGYHUDAICLFK-UHFFFAOYSA-N
tetraoxoosmium
30318
2% aq. soln.
Air Sensitive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P501c

H311-H315-H319-H332

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC LIQUID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 244-058-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only