Learn More
Osmium(VIII) oxide, 2% aq. soln., Thermo Scientific Chemicals
Description
Osmium(VIII) oxide is widely used as an oxidant in organic synthesis to oxidize alkenes to vicinal diols as well as in the sharpless oxyamination to give vicinal amino-alcohols. It is involved in the oxidative cleavage of alkenes (Lemieux-Johnson oxidation) in association with sodium periodate. It finds application in transmission electron microscopy (TEM) as a staining agent. It is involved in the promotion of oxidative cleavage of graphene layers in carbon nanotubes and as an intermediate in osmium ore refining. As a lipid stain, it is useful in scanning electron microscopy (SEM) as an alternative to sputter coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless to Yellow |
Physical Form | Liquid |
Quantity | 10 mL |
UN Number | UN3287 |
Merck Index | 14,6893 |
Solubility Information | Soluble in alcohol,ether,chloroform,benzene,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride |
Formula Weight | 254.1 |
Concentration or Composition (by Analyte or Components) | 2% aq. soln. |
Odor | Chlorine-like |
Sensitivity | Air Sensitive |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y01K032 | Certificate of Analysis | 16/12/2024 | 045384.AB,045384.9W |
T26K033 | Certificate of Analysis | 21/08/2024 | 045384.AB,045384.9W |
Q25K005 | Certificate of Analysis | 11/06/2024 | 045384.AB,045384.9W |
Q29K049 | Certificate of Analysis | 10/05/2024 | 045384.AB,045384.9W |
X16J051 | Certificate of Analysis | 20/10/2023 | 045384.AB,045384.9W |
Safety and Handling

- Osmium(VIII) oxide, 2% aqueous solution
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Osmium(VIII) oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.