Learn More
BINDER™ Overheating Alarm Function for Drying and Heating Chambers
The Overheating Alarm Function for Drying and Heating Chambers features a switchable acoustic alarm, with adjustable limit value on the independent temperature safety device.
Brand:  BINDER™ 8012-1299
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 16434169
  • £680.00 / Each
Estimated Shipment: 16-09-2024
to see stock.
Add to basket
Description
Description
  • Audible alarm function for overheating
  • Features switchable acoustic alarm
  • Features adjustable limit value
  • Designed for use with BINDER™ Drying and Heating Chambers
Specifications
Specifications
Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device
Adjustable Overheating Alarm
230 V
BINDER Chambers ED 23 Drying and Heating Chambers with Natural Convection, 230/400 V Model
Switchable acoustic alarm