missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ Blotting Paper

Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with this high-quality blotting paper.

Brand:  Thermo Scientific™ FP-7

59.47 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.



Code : 04

Additional Details : Weight : 0.55836kg

 View more versions of this product

Product Code. 12444637

  • £47.00 / Pack of 100
Estimated Shipment: 13-11-2024
to see stock.
Add to basket

Description

Description

Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.

  • High purity, 100% cotton fiber
  • Available in pre-cut mini gel and large format gel sizes
  • Compatible with Owl electroblotting systems

Ordering Information:

  • Sold in packs of 100 sheets
Specifications

Specifications

12 cm
Sheets
Owl electroblotting systems
Sheet
12 cm x 16 cm
Owl
16 cm
35 mL/min.
183 g/m²
100% Cotton Fiber
0.33 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers